Product Details
- SNP ID
-
rs3129157
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:29173966 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTACTTTGTTCTTGCACTGGGAATC[A/G]CAGAGTGTGTCCTACTGGTGGTGAT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR2J2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3129158] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR2J2
- Gene Name
- olfactory receptor family 2 subfamily J member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030905.2 |
433 |
Missense Mutation |
ACA,GCA |
T111A |
NP_112167.2 |
View Full Product Details