Product Details

SNP ID
rs41270632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29174570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTATTGGGGTGGGAGTGGGGGA[A/T]GTGACAGGGAAATCATGTTGTCTGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2J2 PubMed Links

Gene Details

Gene
OR2J2
Gene Name
olfactory receptor family 2 subfamily J member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030905.2 1037 Missense Mutation AAG,ATG K312M NP_112167.2

View Full Product Details