Product Details

SNP ID
rs138014515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52834257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGATCATTTCAGTCAAATCTA[C/T]TATACCTTCTGTGTACATATCAATC
Phenotype
MIM: 607605
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTA5 PubMed Links

Gene Details

Gene
GSTA5
Gene Name
glutathione S-transferase alpha 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153699.1 368 Missense Mutation ATA,GTA I100V NP_714543.1

View Full Product Details