Product Details

SNP ID
rs138251788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:123216886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGAAAGAATTAATGGACAAACC[A/G]TTCTGTTCTATGAAATCTAAAACAT
Phenotype
MIM: 603283
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRDN PubMed Links

Gene Details

Gene
TRDN
Gene Name
triadin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251987.1 4223 Intron NP_001238916.1
NM_001256020.1 4223 Intron NP_001242949.1
NM_001256021.1 4223 Intron NP_001242950.1
NM_001256022.1 4223 Intron NP_001242951.1
NM_006073.3 4223 UTR 3 NP_006064.2

View Full Product Details