Product Details

SNP ID
rs138283298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:151366010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCTTCACCTCCCTCTCCTGGTC[A/G]GCCTGCTCCAGGTTGTCAATGTCAG
Phenotype
MIM: 604698 MIM: 616595
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKAP12 PubMed Links

Gene Details

Gene
AKAP12
Gene Name
A-kinase anchoring protein 12
There are no transcripts associated with this gene.

Gene
ZBTB2
Gene Name
zinc finger and BTB domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020861.2 1431 Silent Mutation GCC,GCT A352A NP_065912.1
XM_005267076.2 1431 Silent Mutation GCC,GCT A352A XP_005267133.1
XM_011536004.2 1431 Silent Mutation GCC,GCT A352A XP_011534306.1
XM_017011137.1 1431 Silent Mutation GCC,GCT A273A XP_016866626.1

View Full Product Details