Product Details
- SNP ID
-
rs138317994
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:42765960 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATTTGTTTGTTTGCTACTAATTCC[A/G]GTGCTTTTTGTTTTTGTTTTCTATT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GLTSCR1L
PubMed Links
Gene Details
- Gene
- GLTSCR1L
- Gene Name
- GLTSCR1 like
View Full Product Details