Product Details

SNP ID
rs139287158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29374582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGGGAGGCACAAGTGGACAGAG[A/C]CTTGTGGAGTATTCTGCAGGACCTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR12D3 PubMed Links

Gene Details

Gene
OR12D3
Gene Name
olfactory receptor family 12 subfamily D member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030959.2 710 Missense Mutation GCT,TCT A236S NP_112221.1
Gene
OR5V1
Gene Name
olfactory receptor family 5 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030876.5 710 Intron NP_110503.3
XM_011514929.2 710 Intron XP_011513231.1
XM_011514930.2 710 Intron XP_011513232.1

View Full Product Details