Product Details

SNP ID
rs139321881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41190227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTCTCCTCTCTCACTCCTTCTC[C/T]AGATGAGGAAACAAGATCAGAGAGG
Phenotype
MIM: 609715
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TREML2 PubMed Links

Gene Details

Gene
TREML2
Gene Name
triggering receptor expressed on myeloid cells like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024807.3 3070 UTR 3 NP_079083.2
XM_011514917.2 3070 UTR 3 XP_011513219.1

View Full Product Details