Product Details

SNP ID
rs139512248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:167000109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGCACTAGAGGAGCAAGAAAAAG[G/T]AGAGGTATGAAGTTTCCAGATTTTA
Phenotype
MIM: 605392
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FGFR1OP PubMed Links

Gene Details

Gene
FGFR1OP
Gene Name
FGFR1 oncogene partner
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278690.1 248 Missense Mutation GGA,GTA G51V NP_001265619.1
NM_007045.3 248 Missense Mutation GGA,GTA G51V NP_008976.1
NM_194429.2 248 Missense Mutation GGA,GTA G51V NP_919410.1
Gene
MIR3939
Gene Name
microRNA 3939
There are no transcripts associated with this gene.

View Full Product Details