Product Details

SNP ID
rs139823445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:82365111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTCGGCGCCGTTCCTGGCTTC[C/T]GCCGTGTCCGCCCAGCCCCCGCTGC
Phenotype
MIM: 190920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TPBG PubMed Links

Gene Details

Gene
TPBG
Gene Name
trophoblast glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166392.1 575 Silent Mutation TCC,TCT S50S NP_001159864.1
NM_006670.4 575 Silent Mutation TCC,TCT S50S NP_006661.1
XM_011536097.2 575 Silent Mutation TCC,TCT S50S XP_011534399.1

View Full Product Details