Product Details

SNP ID
rs140320629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:7593765 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTGATATGTCCATACGATTCCA[A/G]TCATCACATGCCTAAATCATCTTTG
Phenotype
MIM: 125647
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DSP PubMed Links

Gene Details

Gene
DSP
Gene Name
desmoplakin
There are no transcripts associated with this gene.

Gene
SNRNP48
Gene Name
small nuclear ribonucleoprotein U11/U12 subunit 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152551.3 247 Missense Mutation AAT,AGT N63S NP_689764.3
XM_011514312.2 247 Missense Mutation AAT,AGT N42S XP_011512614.1

View Full Product Details