Product Details

SNP ID
rs140525877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33319129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCGTGGAGGAATCAGCAACTGG[G/T]GCCAAGGAAGCCAAGGGGGAAGGTG
Phenotype
MIM: 603186 MIM: 601962 MIM: 611439
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DAXX PubMed Links

Gene Details

Gene
DAXX
Gene Name
death domain associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001141969.1 1085 Silent Mutation GCA,GCC A677A NP_001135441.1
NM_001141970.1 1085 Silent Mutation GCA,GCC A689A NP_001135442.1
NM_001254717.1 1085 Silent Mutation GCA,GCC A602A NP_001241646.1
NM_001350.4 1085 Silent Mutation GCA,GCC A677A NP_001341.1
XM_005248860.3 1085 Silent Mutation GCA,GCC A309A XP_005248917.1
Gene
TAPBP
Gene Name
TAP binding protein (tapasin)
There are no transcripts associated with this gene.

Gene
ZBTB22
Gene Name
zinc finger and BTB domain containing 22
There are no transcripts associated with this gene.

View Full Product Details