Product Details

SNP ID
rs141299898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116645894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTATTAGTAAGCATAATGTTCG[G/T]TTTTTTTTCTCTTCAATTCCAATAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZUFSP PubMed Links

Gene Details

Gene
ZUFSP
Gene Name
zinc finger with UFM1 specific peptidase domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145062.2 2162 Silent Mutation NP_659499.2
XM_011535565.2 2162 Silent Mutation XP_011533867.1
XM_011535567.1 2162 Silent Mutation XP_011533869.1
XM_017010393.1 2162 Silent Mutation XP_016865882.1
XM_017010394.1 2162 Missense Mutation XP_016865883.1
XM_017010395.1 2162 Silent Mutation XP_016865884.1
XM_017010396.1 2162 Silent Mutation XP_016865885.1

View Full Product Details