Product Details

SNP ID
rs141334348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33455135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACACCTTTGCCGCCTGTACCCG[C/T]CTCCCCGACCTGCAACCCAGCCCCA
Phenotype
MIM: 603384
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SYNGAP1 PubMed Links

Gene Details

Gene
SYNGAP1
Gene Name
synaptic Ras GTPase activating protein 1
There are no transcripts associated with this gene.

Gene
ZBTB9
Gene Name
zinc finger and BTB domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152735.3 303 Missense Mutation GCC,GTC A12V NP_689948.1

View Full Product Details