Product Details

SNP ID
rs141609699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39105465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCAGGGGTCTCAACTGTAACTC[A/G]CGCTCCAACTGCTCTGCAGTCAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAYSD1 PubMed Links

Gene Details

Gene
SAYSD1
Gene Name
SAYSVFN motif domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304793.1 616 Silent Mutation CGC,CGT R106R NP_001291722.1
NM_018322.2 616 Silent Mutation CGC,CGT R173R NP_060792.1

View Full Product Details