Product Details

SNP ID
rs141689063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26199194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAGTCCGGCCCGCGAAGAGCGG[A/G]TCTTAGCCTTAGCTCGGGCCTTTCC
Phenotype
MIM: 602792 MIM: 602804 MIM: 602811 MIM: 602830
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIST1H2AD PubMed Links

Gene Details

Gene
HIST1H2AD
Gene Name
histone cluster 1, H2ad
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021065.3 100 Missense Mutation NP_066409.1
Gene
HIST1H2BF
Gene Name
histone cluster 1, H2bf
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003522.3 100 Intron NP_003513.1
Gene
HIST1H3D
Gene Name
histone cluster 1, H3d
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003530.4 100 UTR 5 NP_003521.2
Gene
HIST1H4E
Gene Name
histone cluster 1, H4e
There are no transcripts associated with this gene.

View Full Product Details