Product Details

SNP ID
rs141968346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:75253002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGATTTTTATGTTTACTGAATGG[A/T]GCATGGGATAAAAAGGGGTTCTAAA
Phenotype
MIM: 123996 MIM: 611028
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
COX7A2 PubMed Links

Gene Details

Gene
COX7A2
Gene Name
cytochrome c oxidase subunit 7A2
There are no transcripts associated with this gene.

Gene
TMEM30A
Gene Name
transmembrane protein 30A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143958.1 4356 UTR 3 NP_001137430.1
NM_018247.3 4356 UTR 3 NP_060717.1

View Full Product Details