Product Details

SNP ID
rs141980901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35499990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTCAGGTACTCACGGTCATCAG[C/T]GTGGACAATCTGGAAGTTCTTGACT
Phenotype
MIM: 603170 MIM: 602280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEAD3 PubMed Links

Gene Details

Gene
TEAD3
Gene Name
TEA domain transcription factor 3
There are no transcripts associated with this gene.

Gene
TULP1
Gene Name
tubby like protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289395.1 1530 Missense Mutation ACT,GCT T443A NP_001276324.1
NM_003322.4 1530 Missense Mutation ACT,GCT T496A NP_003313.3

View Full Product Details