Product Details

SNP ID
rs142486086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:133889424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCACCGGCTCCCTCAGCGATGT[A/G]GAGGACCTTCAAGAGGTGGAGATGT
Phenotype
MIM: 616058 MIM: 603306
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TARID PubMed Links

Gene Details

Gene
TARID
Gene Name
TCF21 antisense RNA inducing promoter demethylation
There are no transcripts associated with this gene.

Gene
TCF21
Gene Name
transcription factor 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003206.3 304 Silent Mutation GTA,GTG V9V NP_003197.2
NM_198392.2 304 Silent Mutation GTA,GTG V9V NP_938206.1

View Full Product Details