Product Details

SNP ID
rs142499546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:8413546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTATTTTAAATAGGACAATCAC[C/T]GTCTATACAGTCTGTGCCAGCGTCC
Phenotype
MIM: 610845
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC35B3 PubMed Links

Gene Details

Gene
SLC35B3
Gene Name
solute carrier family 35 member B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142541.2 1430 UTR 3 NP_001136013.1
NM_015948.4 1430 UTR 3 NP_057032.2
XM_005249156.3 1430 UTR 3 XP_005249213.1
XM_005249157.2 1430 UTR 3 XP_005249214.1
XM_005249158.1 1430 UTR 3 XP_005249215.1
XM_006715102.3 1430 UTR 3 XP_006715165.1
XM_006715103.3 1430 UTR 3 XP_006715166.1
XM_011514648.1 1430 UTR 3 XP_011512950.1
XM_011514650.2 1430 Intron XP_011512952.1
XM_017010910.1 1430 UTR 3 XP_016866399.1
XM_017010911.1 1430 UTR 3 XP_016866400.1
XM_017010912.1 1430 UTR 3 XP_016866401.1
XM_017010913.1 1430 UTR 3 XP_016866402.1
XM_017010914.1 1430 UTR 3 XP_016866403.1
XM_017010915.1 1430 UTR 3 XP_016866404.1

View Full Product Details