Product Details

SNP ID
rs142668485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:40391715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCGCCTTTCCAAACACTCAGGG[C/T]TCAGTCCGGCATTTGAGCGAGTCCA
Phenotype
MIM: 612808
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRFN2 PubMed Links

Gene Details

Gene
LRFN2
Gene Name
leucine rich repeat and fibronectin type III domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020737.2 4009 UTR 3 NP_065788.1
XM_011514761.2 4009 UTR 3 XP_011513063.1
XM_011514762.2 4009 UTR 3 XP_011513064.1
XM_017011110.1 4009 UTR 3 XP_016866599.1
XM_017011111.1 4009 Intron XP_016866600.1

View Full Product Details