Product Details

SNP ID
rs142669991
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33276186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCTGAATCCAGGTCCTAGCCA[A/G]TGGTCTGGACAACAAGCTCCGTGAA
Phenotype
MIM: 603095 MIM: 180473 MIM: 603443 MIM: 611440
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
B3GALT4 PubMed Links
Additional Information
For this assay, SNP(s) [rs72654480] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
B3GALT4
Gene Name
beta-1,3-galactosyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003782.3 Intron NP_003773.1
Gene
RPS18
Gene Name
ribosomal protein S18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022551.2 Intron NP_072045.1
Gene
VPS52
Gene Name
VPS52, GARP complex subunit
There are no transcripts associated with this gene.

Gene
WDR46
Gene Name
WD repeat domain 46
There are no transcripts associated with this gene.

View Full Product Details