Product Details

SNP ID
rs142998748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169702870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGAGATGAAGGTGTACTACGACGG[C/G]ACGGTCGAGCAGCACCCGTTCGGCG
Phenotype
MIM: 616987 MIM: 613069
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C6orf120 PubMed Links

Gene Details

Gene
C6orf120
Gene Name
chromosome 6 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029863.2 759 Silent Mutation GGC,GGG G137G NP_001025034.1
NM_001317342.1 759 Silent Mutation GGC,GGG G156G NP_001304271.1
Gene
PHF10
Gene Name
PHD finger protein 10
There are no transcripts associated with this gene.

Gene
WDR27
Gene Name
WD repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202550.1 759 Intron NP_001189479.1
NM_182552.4 759 Intron NP_872358.4
XM_011535682.2 759 Intron XP_011533984.1
XM_011535684.2 759 Intron XP_011533986.1
XM_011535685.2 759 Intron XP_011533987.1
XM_011535687.2 759 Intron XP_011533989.1
XM_011535688.2 759 Intron XP_011533990.1
XM_011535689.2 759 Intron XP_011533991.1
XM_011535690.2 759 Intron XP_011533992.1
XM_011535691.2 759 Intron XP_011533993.1
XM_011535692.2 759 Intron XP_011533994.1
XM_011535693.2 759 Intron XP_011533995.1
XM_011535694.2 759 Intron XP_011533996.1
XM_011535696.2 759 Intron XP_011533998.1
XM_011535697.2 759 Intron XP_011533999.1
XM_017010658.1 759 Intron XP_016866147.1
XM_017010659.1 759 Intron XP_016866148.1
XM_017010660.1 759 Intron XP_016866149.1
XM_017010661.1 759 Intron XP_016866150.1
XM_017010662.1 759 Intron XP_016866151.1
XM_017010663.1 759 Intron XP_016866152.1
XM_017010664.1 759 Intron XP_016866153.1
XM_017010665.1 759 Intron XP_016866154.1
XM_017010666.1 759 Intron XP_016866155.1
XM_017010667.1 759 Intron XP_016866156.1
XM_017010668.1 759 Intron XP_016866157.1
XM_017010669.1 759 Intron XP_016866158.1
XM_017010670.1 759 Intron XP_016866159.1
XM_017010671.1 759 Intron XP_016866160.1
XM_017010672.1 759 Intron XP_016866161.1
XM_017010673.1 759 Intron XP_016866162.1
XM_017010674.1 759 Intron XP_016866163.1
XM_017010675.1 759 Intron XP_016866164.1

View Full Product Details