Product Details

SNP ID
rs143228836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30723410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCGCCGAGCTGGTTGATTCTGT[C/T]CTGGATGTGGTACGGAAGGAGGCAG
Phenotype
MIM: 606998 MIM: 607593 MIM: 191130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLOT1 PubMed Links

Gene Details

Gene
FLOT1
Gene Name
flotillin 1
There are no transcripts associated with this gene.

Gene
MDC1
Gene Name
mediator of DNA damage checkpoint 1
There are no transcripts associated with this gene.

Gene
TUBB
Gene Name
tubulin beta class I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293212.1 738 Silent Mutation GTC,GTT V136V NP_001280141.1
NM_001293213.1 738 Silent Mutation GTC,GTT V116V NP_001280142.1
NM_001293214.1 738 Silent Mutation GTC,GTT V72V NP_001280143.1
NM_001293215.1 738 Silent Mutation GTC,GTT V44V NP_001280144.1
NM_001293216.1 738 Silent Mutation GTC,GTT V44V NP_001280145.1
NM_178014.3 738 Silent Mutation GTC,GTT V116V NP_821133.1

View Full Product Details