Product Details

SNP ID
rs143578587
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169705201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTCACACATATCACAGAACATC[A/G]TTTCTTCTTCATGGTGGGGTTGTCC
Phenotype
MIM: 616987 MIM: 613069
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C6orf120 PubMed Links

Gene Details

Gene
C6orf120
Gene Name
chromosome 6 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029863.2 3090 UTR 3 NP_001025034.1
NM_001317342.1 3090 UTR 3 NP_001304271.1
Gene
PHF10
Gene Name
PHD finger protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018288.3 3090 Missense Mutation ACG,ATG T448M NP_060758.2
NM_133325.2 3090 Missense Mutation ACG,ATG T446M NP_579866.2
XM_017010998.1 3090 Missense Mutation ACG,ATG T396M XP_016866487.1
Gene
WDR27
Gene Name
WD repeat domain 27
There are no transcripts associated with this gene.

View Full Product Details