Product Details

SNP ID
rs143979349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:83211980 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCTTCAGGCCAAGAATAACAAT[C/T]AGGTAGAATCTGGTCATAATCAGTA
Phenotype
MIM: 154250
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ME1 PubMed Links

Gene Details

Gene
ME1
Gene Name
malic enzyme 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002395.5 1928 Missense Mutation AAT,GAT N555D NP_002386.1
XM_011535836.2 1928 Missense Mutation AAT,GAT N480D XP_011534138.1

View Full Product Details