Product Details

SNP ID
rs144181034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27451894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATTCATGACACTGATAGGGTTT[A/C]TCTCCAGTATGAATTCTTTCATGCT
Phenotype
MIM: 602277
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF184 PubMed Links

Gene Details

Gene
ZNF184
Gene Name
zinc finger protein 184
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318891.1 1950 Missense Mutation GAG,GAT E555D NP_001305820.1
NM_001318892.1 1950 Missense Mutation GAG,GAT E555D NP_001305821.1
NM_001318893.1 1950 Missense Mutation GAG,GAT E479D NP_001305822.1
NM_007149.2 1950 Missense Mutation GAG,GAT E555D NP_009080.2
XM_011514861.2 1950 Missense Mutation GAG,GAT E479D XP_011513163.1

View Full Product Details