Product Details

SNP ID
rs144318583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28259414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAGCTATTCCGAGGACATCGTG[G/T]GCTCTCGGAGAAGGCGACGCAGCTC
Phenotype
MIM: 611643 MIM: 616474
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NKAPL PubMed Links

Gene Details

Gene
NKAPL
Gene Name
NFKB activating protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007531.2 118 Missense Mutation GGC,TGC G15C NP_001007532.1
Gene
ZKSCAN4
Gene Name
zinc finger with KRAB and SCAN domains 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304506.1 118 Intron NP_001291435.1
NM_019110.4 118 Intron NP_061983.2
XM_005249095.3 118 Intron XP_005249152.1
XM_005249097.3 118 Intron XP_005249154.1
XM_005249098.3 118 Intron XP_005249155.1
XM_005249100.4 118 Intron XP_005249157.2
XM_006715095.3 118 Intron XP_006715158.1
XM_011514588.2 118 Intron XP_011512890.1
XM_011514589.2 118 Intron XP_011512891.1
XM_017010844.1 118 Intron XP_016866333.1
Gene
ZSCAN26
Gene Name
zinc finger and SCAN domain containing 26
There are no transcripts associated with this gene.

View Full Product Details