Product Details

SNP ID
rs144557524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30489839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCAGTTCGTGCGCTTCGACAAC[G/T]ACGCCGCGAGTCCGAGGATGGTGCC
Phenotype
MIM: 143010
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HLA-E PubMed Links

Gene Details

Gene
HLA-E
Gene Name
major histocompatibility complex, class I, E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005516.5 316 Missense Mutation GAC,TAC D60Y NP_005507.3
XM_017010807.1 316 Missense Mutation GAC,TAC D101Y XP_016866296.1
XM_017010808.1 316 Missense Mutation GAC,TAC D101Y XP_016866297.1
XM_017010809.1 316 Missense Mutation GAC,TAC D60Y XP_016866298.1

View Full Product Details