Product Details
- SNP ID
-
rs145158912
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:38682101 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCAGGAACAGCAATTCCAATATGA[A/C]CTTACGTGATACCCCCCGAAAAAAG
- Phenotype
-
MIM: 138750
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GLO1
PubMed Links
Gene Details
- Gene
- GLO1
- Gene Name
- glyoxalase I
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006708.2 |
499 |
Missense Mutation |
|
|
NP_006699.2 |
View Full Product Details