Product Details

SNP ID
rs145355467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:167179047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTCCCGAAGTCTGCAAAACTCC[C/G]GTGGCAGAAAATCACCAGTGCAGGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC105378123 PubMed Links

Gene Details

Gene
LOC105378123
Gene Name
uncharacterized LOC105378123
There are no transcripts associated with this gene.

Gene
TCP10L2
Gene Name
t-complex 10-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145121.1 Intron NP_001138593.1
XM_017010859.1 Intron XP_016866348.1

View Full Product Details