Product Details

SNP ID
rs145827595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43018708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCGGCACTTCCATGACCTCTGGA[A/G]GTTTAATCCTGGTAAAGAGCACTGC
Phenotype
MIM: 611248 MIM: 143170 MIM: 601646 MIM: 613475
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHDC3 PubMed Links

Gene Details

Gene
KLHDC3
Gene Name
kelch domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_057161.3 1114 Missense Mutation AAG,AGG K270R NP_476502.1
Gene
MEA1
Gene Name
male-enhanced antigen 1
There are no transcripts associated with this gene.

Gene
PPP2R5D
Gene Name
protein phosphatase 2 regulatory subunit B'delta
There are no transcripts associated with this gene.

Gene
RRP36
Gene Name
ribosomal RNA processing 36
There are no transcripts associated with this gene.

View Full Product Details