Product Details

SNP ID
rs145923049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:87085795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAACAAGTACTGCAGTGGCACGC[C/T]GTGTGGTTCTCCACTTTGAAACCCC
Phenotype
MIM: 118850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CGA PubMed Links

Gene Details

Gene
CGA
Gene Name
glycoprotein hormones, alpha polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000735.3 547 Silent Mutation ACA,ACG T104T NP_000726.1
NM_001252383.1 547 Silent Mutation ACA,ACG T135T NP_001239312.1

View Full Product Details