Product Details

SNP ID
rs145957272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31763521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAAGACCATCTGAGGGAGGCAC[A/G]GCGACAGCAGCTGCATCTAGGGGCC
Phenotype
MIM: 603382 MIM: 609693
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MSH5 PubMed Links

Gene Details

Gene
MSH5
Gene Name
mutS homolog 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002441.4 280 Intron NP_002432.1
NM_025259.5 280 Intron NP_079535.4
NM_172165.3 280 Intron NP_751897.1
NM_172166.3 280 Intron NP_751898.1
Gene
MSH5-SAPCD1
Gene Name
MSH5-SAPCD1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
SAPCD1
Gene Name
suppressor APC domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039651.1 280 Missense Mutation CAG,CGG Q74R NP_001034740.1
Gene
SAPCD1-AS1
Gene Name
SAPCD1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
VWA7
Gene Name
von Willebrand factor A domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025258.2 280 Intron NP_079534.2
XM_005249427.2 280 Intron XP_005249484.1
XM_017011327.1 280 Intron XP_016866816.1
XM_017011328.1 280 Intron XP_016866817.1
XM_017011329.1 280 Intron XP_016866818.1

View Full Product Details