Product Details

SNP ID
rs80001134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31972121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGACACCAACGTGGCCACCGGCG[C/T]CCCTCCACGCCGCCAACGAGTCCCC
Phenotype
MIM: 120810 MIM: 605996 MIM: 600478 MIM: 604977
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C4A PubMed Links

Gene Details

Gene
C4A
Gene Name
complement component 4A (Rodgers blood group)
There are no transcripts associated with this gene.

Gene
DXO
Gene Name
decapping exoribonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005510.3 253 UTR 5 NP_005501.2
XM_006715005.3 253 UTR 5 XP_006715068.1
XM_017010329.1 253 UTR 5 XP_016865818.1
Gene
SKIV2L
Gene Name
Ski2 like RNA helicase
There are no transcripts associated with this gene.

Gene
STK19
Gene Name
serine/threonine kinase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004197.1 253 Missense Mutation GCC,GTC A42V NP_004188.1
NM_032454.1 253 Missense Mutation GCC,GTC A42V NP_115830.1

View Full Product Details