Product Details

SNP ID
rs146273799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26251675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCCTGATCCAGCTAAGTCCGCT[C/G]CCGCCCCGAAGAAGGGCTCCAAGAA
Phenotype
MIM: 602806 MIM: 602816 MIM: 602832
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HIST1H2BH PubMed Links

Gene Details

Gene
HIST1H2BH
Gene Name
histone cluster 1, H2bh
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003524.2 Intron NP_003515.1
Gene
HIST1H3F
Gene Name
histone cluster 1, H3f
There are no transcripts associated with this gene.

Gene
HIST1H4G
Gene Name
histone cluster 1, H4g
There are no transcripts associated with this gene.

View Full Product Details