Product Details

SNP ID
rs146275920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42963074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGCCCGTGCCTGGAGCTCAGGG[A/G]ACCAGAGTGAGCACCGGCTGGCGCT
Phenotype
MIM: 606628 MIM: 601498
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNPY3-GNMT PubMed Links

Gene Details

Gene
CNPY3-GNMT
Gene Name
CNPY3-GNMT readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318856.1 468 Missense Mutation AAC,GAC N86D NP_001305785.1
NM_001318857.1 468 Missense Mutation AAC,GAC N91D NP_001305786.1
NM_001318858.1 468 Intron NP_001305787.1
Gene
GNMT
Gene Name
glycine N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318865.1 468 Intron NP_001305794.1
NM_018960.5 468 Missense Mutation AAC,GAC N152D NP_061833.1
Gene
PEX6
Gene Name
peroxisomal biogenesis factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000287.3 468 Intron NP_000278.3
NM_001316313.1 468 Intron NP_001303242.1
XM_011514661.1 468 Intron XP_011512963.1

View Full Product Details