Product Details

SNP ID
rs146572999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:127578706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGCAGATTAGCTGATCCAACC[A/C]GAAGGACAAACTGTGGCTATGAATC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C6orf58 PubMed Links

Gene Details

Gene
C6orf58
Gene Name
chromosome 6 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010905.2 334 Silent Mutation AGA,CGA R108R NP_001010905.1

View Full Product Details