Product Details

SNP ID
rs146633455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:86941016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGAGTGTGCCGTCACAAGTGAC[A/G]AGGGGACAGATGAACAGGGAAGAGA
Phenotype
MIM: 182132
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HTR1E PubMed Links

Gene Details

Gene
HTR1E
Gene Name
5-hydroxytryptamine receptor 1E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000865.2 Intron NP_000856.1
XM_011535789.2 Intron XP_011534091.1

View Full Product Details