Product Details

SNP ID
rs146717555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31960237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCAGAAAATGGCAGAGGAAGA[C/T]GGATCCCTGGTCTCTTTTGGCTGTC
Phenotype
MIM: 138470 MIM: 605996 MIM: 154040 MIM: 600478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFB PubMed Links

Gene Details

Gene
CFB
Gene Name
complement factor B
There are no transcripts associated with this gene.

Gene
DXO
Gene Name
decapping exoribonuclease
There are no transcripts associated with this gene.

Gene
MIR1236
Gene Name
microRNA 1236
There are no transcripts associated with this gene.

Gene
NELFE
Gene Name
negative elongation factor complex member E
There are no transcripts associated with this gene.

Gene
SKIV2L
Gene Name
Ski2 like RNA helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006929.4 643 Missense Mutation ACG,ATG T85M NP_008860.4
XM_011514815.2 643 Missense Mutation ACG,ATG T85M XP_011513117.1

View Full Product Details