Product Details

SNP ID
rs147244701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49606885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATCATCATAGCAGTTCTGGTCA[G/T]ATGGCTGTCCCCAGAGAGGCAACTT
Phenotype
MIM: 180297
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RHAG PubMed Links

Gene Details

Gene
RHAG
Gene Name
Rh-associated glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000324.2 1237 Missense Mutation TAT,TCT Y392S NP_000315.2
XM_011514788.1 1237 Intron XP_011513090.1

View Full Product Details