Product Details
- SNP ID
-
rs147348343
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:34242723 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTGTCTTTACAGAAGGAGCCCAG[C/T]GAAGTGCCAACACCTAAGAGACCTC
- Phenotype
-
MIM: 611419
MIM: 600701
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C6orf1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139876191] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C6orf1
- Gene Name
- chromosome 6 open reading frame 1
There are no transcripts associated with this gene.
- Gene
- HMGA1
- Gene Name
- high mobility group AT-hook 1
- Gene
- MIR6835
- Gene Name
- microRNA 6835
There are no transcripts associated with this gene.
View Full Product Details