Product Details

SNP ID
rs147634249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49728781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGCCTTGCAACTGTCCCTGAC[C/T]AACTGATGTTTACAGGTTAATGTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRISP3 PubMed Links

Gene Details

Gene
CRISP3
Gene Name
cysteine rich secretory protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190986.1 814 Silent Mutation TTA,TTG L252L NP_001177915.1
NM_006061.2 814 Silent Mutation TTA,TTG L242L NP_006052.2

View Full Product Details