Product Details

SNP ID
rs147713380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43650423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGCCCCCTGCCACAGAGGAGAT[A/G]GTGGCGCAGTCGTCTGTGGTGAAGG
Phenotype
MIM: 612648
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAD2L1BP PubMed Links

Gene Details

Gene
MAD2L1BP
Gene Name
MAD2L1 binding protein
There are no transcripts associated with this gene.

Gene
RSPH9
Gene Name
radial spoke head 9 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193341.1 345 Missense Mutation ATA,ATG I92M NP_001180270.1
NM_152732.4 345 Missense Mutation ATA,ATG I92M NP_689945.2
XM_005248901.3 345 Missense Mutation ATA,ATG I92M XP_005248958.1
XM_006715014.1 345 Intron XP_006715077.1
XM_017010426.1 345 Missense Mutation ATA,ATG I92M XP_016865915.1

View Full Product Details