Product Details

SNP ID
rs148607776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41905793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGCTCAGGAACCATGAAGCAT[A/G]GTGCAGGTATCAGGGTCAAGTATTC
Phenotype
MIM: 612915
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED20 PubMed Links

Gene Details

Gene
MED20
Gene Name
mediator complex subunit 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305455.1 2164 UTR 3 NP_001292384.1
NM_001305456.1 2164 UTR 3 NP_001292385.1
NM_001305457.1 2164 UTR 3 NP_001292386.1
NM_004275.4 2164 UTR 3 NP_004266.2

View Full Product Details