Product Details

SNP ID
rs148698842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:107068740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTTTGCTTTCTTGAGGATGTC[A/G]CATTTTTTCCTGTTGGGGCGGCGGC
Phenotype
MIM: 616374
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BEND3 PubMed Links

Gene Details

Gene
BEND3
Gene Name
BEN domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080450.2 3101 Silent Mutation TGC,TGT C817C NP_001073919.1
XM_005267079.3 3101 Intron XP_005267136.1
XM_005267080.3 3101 Silent Mutation TGC,TGT C817C XP_005267137.1
XM_011536005.2 3101 Intron XP_011534307.1
XM_017011138.1 3101 Silent Mutation TGC,TGT C817C XP_016866627.1

View Full Product Details