Product Details

SNP ID
rs148742612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:97146848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTCTCTGCATCTCATCTTGTCC[C/T]ATCTGTCCAAGGTGAGACAAAAGTT
Phenotype
MIM: 615614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL32 PubMed Links

Gene Details

Gene
KLHL32
Gene Name
kelch like family member 32
There are no transcripts associated with this gene.

Gene
MMS22L
Gene Name
MMS22 like, DNA repair protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198468.2 3956 Missense Mutation ATA,ATG I1230M NP_940870.2
XM_006715430.1 3956 Missense Mutation ATA,ATG I1230M XP_006715493.1
XM_006715432.3 3956 Intron XP_006715495.1
XM_011535670.2 3956 Intron XP_011533972.1
XM_011535671.2 3956 Intron XP_011533973.1
XM_011535672.2 3956 Intron XP_011533974.1
XM_011535673.2 3956 Intron XP_011533975.1
XM_011535674.2 3956 Intron XP_011533976.1
XM_011535675.2 3956 Intron XP_011533977.1
XM_011535676.2 3956 Intron XP_011533978.1
XM_011535677.2 3956 Intron XP_011533979.1
XM_011535678.2 3956 Intron XP_011533980.1
XM_011535679.2 3956 Intron XP_011533981.1
XM_011535680.2 3956 Intron XP_011533982.1
XM_017010656.1 3956 Intron XP_016866145.1
XM_017010657.1 3956 Missense Mutation ATA,ATG I947M XP_016866146.1

View Full Product Details