Product Details

SNP ID
rs148814957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26216641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGCTGACTTAGCTGGTTCAGG[A/C]ATGCTGTCAGAAAACAATAACAGCA
Phenotype
MIM: 602786 MIM: 602798 MIM: 602813
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HIST1H2AE PubMed Links

Gene Details

Gene
HIST1H2AE
Gene Name
histone cluster 1, H2ae
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021052.2 Intron NP_066390.1
Gene
HIST1H2BG
Gene Name
histone cluster 1, H2bg
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003518.3 Intron NP_003509.1
Gene
HIST1H3E
Gene Name
histone cluster 1, H3e
There are no transcripts associated with this gene.

View Full Product Details