Product Details

SNP ID
rs148820897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27912077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGTGTTGCATATGTTTACCAG[C/T]ATTTGTGGAACTGTACTTGTGGTAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2B2 PubMed Links

Gene Details

Gene
OR2B2
Gene Name
olfactory receptor family 2 subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033057.2 320 Missense Mutation ATA,ATG I81M NP_149046.2

View Full Product Details