Product Details

SNP ID
rs148879002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36954832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGCTGGTGGCCACCACAGGCCC[C/T]GTTGGAGCCCTCACAGATGAGGAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PI16 PubMed Links

Gene Details

Gene
PI16
Gene Name
peptidase inhibitor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199159.1 295 Silent Mutation CCC,CCT P24P NP_001186088.1
NM_153370.2 295 Silent Mutation CCC,CCT P24P NP_699201.2
XM_005248917.2 295 Silent Mutation CCC,CCT P24P XP_005248974.1
XM_011514375.2 295 Silent Mutation CCC,CCT P24P XP_011512677.1
XM_017010430.1 295 Silent Mutation CCC,CCT P24P XP_016865919.1

View Full Product Details